Learn More
Essendant BUFFERFLOORPAD16RD
Supplier:  Essendant MMM08391
Encompass_Preferred
3M Red Buffer Pads 5100 can be used for everyday cleaning remove light soil scuff marks and black heel marks Its regular use reduces the frequency of scrub-and-recoat operations enhancing the appearance of your finished floors with reduced labor and materials costs Fibers throughout the pad are uniformly coated with a high-quality resin and fine cleaning particles providing good performance throughout the pad s long life Open-web construction means this washable reusable pad resists clogging and loading Conformable fibers provide maximum pad-to-floor contact Pads are reversible For rotary swing machines and automatic floor machines running at low speed (175-600 rpm) Floor Pads Application- Buffing Diameter- 16 Color(s)- Red Color Family- Red
Catalog No. 50-000-32949