Learn More
Medchemexpress LLC Buflomedil (hydrochloride) | 35543-24-9 | 99.8% | 10 G

Supplier: Medchemexpress LLC HYB048410G
Buflomedil hydrochloride is a vasodilator agent and an orally active α1A-adrenoceptor antagonist. It is primarily used for the study of peripheral circulatory disorders and exhibits specific Ki values for α1A-AR and α1B-AR.
- Vasodilator agent
- Orally active α1A-adrenoceptor antagonist
- Used in the study of peripheral circulatory disorders
- Ki of 4.06 μM for rat α1A-adrenergic receptor
- Ki of 6.84 μM for α1B-adrenergic receptor
- Purity of 99.8%
- Molecular formula: C17H26ClNO4
- Molecular weight: 343.85
- Appearance: white to off-white solid
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.