Learn More
Lonza Walkersville AGM Astrocyte Growth Medium BulletKit
Supplier:  Lonza Walkersville CC3186
Encompass
Clonetics AGM Astrocyte Growth Medium has been developed to support the growth of primary astrocytes. Media products are provided as convenient BulletKits which contain basal medium and necessary growth factors for cell culture.Media are supplied in several formats:Basal Medium - Contains no growth factors cytokines, or supplementsSingleQuots - Growth factors, cytokines, and supplements (basel media not included)BulletKits- Basal media and SingleQuots Kit packaged togetherContent:1 x ABM Basal Medium (CC-3187), 500 mL1 x AGM SingleQuots Supplement Pack (CC-4123) containing:1 x Bottle FBS, 15.0 mL1 x Bottle L-Glutamine, 5.00 mL1 x Vial GA-1000, 0.50 mL1 x Vial Ascorbic Acid, 0.50 mL1 x Vial HEGF, 0.50 mL1 x Vial Insulin, 1.25 mL
Catalog No. NC9271925