Learn More
Selleck Chemical LLC Bumetanide S1287-1g
SDP
Supplier:  Selleck Chemical LLC S12871G
Encompass_Preferred
Bumetanide is a loop diuretic of the sulfamyl category to treat heart failure Bumetanide is a selective Na -K -Cl- cotransporter 1 (NKCC1) inhibitor weakly inhibits NKCC2 with IC50s of 0 68 and 4 0 M for hNKCC1A and hNKCC2A respectively
Catalog No. 50-311-0638