Learn More
Medchemexpress LLC Bumetanide | 28395-03-1 | 99.97% | 364.42 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY17468R
Encompass_Preferred
Bumetanide (Standard) is the analytical standard of Bumetanide, intended for research and analytical applications. It is a highly potent loop diuretic and a Na+-K+-Cl+ cotransporter (NKCC) blocker. Bumetanide selectively inhibits NKCC1, and also inhibits NKCC2. This compound is used as a reference standard in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard for research and analytical applications.
  • Highly potent loop diuretic.
  • Na+-K+-Cl+ cotransporter (NKCC) blocker.
  • Selective NKCC1 inhibitor, also inhibits NKCC2.
  • Used in HPLC, GC, and MS research experiments.
Catalog No. 50-003-84281