Learn More
Medchemexpress LLC Bumetanide-d5 | 1216739-35-3 | 99.8% | 369.45 | 500 UG
SDP
Supplier:  Medchemexpress LLC HY17468S500UG
Encompass_Preferred
Bumetanide-d5 is a deuterium labeled Bumetanide. It is a selective Na+-K+-Cl- (NKCC1) inhibitor, weakly inhibiting NKCC2 with IC50s of 0.68 and 4.0 μM for hNKCC1A and hNKCC2A, respectively.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-86436