Learn More
Grainger Bumper 1-3/32 in Black Rubber PK10
Supplier:  Grainger 16ZD63
Encompass_Preferred
The Grainger Approved Bumper is a black rubber bumper that provides impact absorption and noise reduction. It measures 4" wide x 2" high x 2" deep, weighs 0.31 lbs, and has a temperature range of -20°F to 150°F. This bumper has a shore A durometer hardness of 60A. It can be used on material handling equipment, grocery carts, hospital beds, and more to help protect walls, racks, and other equipment. Made in the USA.
Catalog No. 50-276-7865