Learn More
Grainger Bumper 3/4 in Black Rubber PK25
Supplier:  Grainger 16XU87
Encompass_Preferred
The Grainger Approved 3/4 in Black Rubber Bumper is a rubber bumper designed to help absorb shock and reduce noise from impacts. It has a 3/4 inch diameter and is made from durable black rubber. This bumper can be used in a variety of applications including on doors, cabinets, and machinery. It has pre-drilled holes for easy mounting and is sold individually. The overall dimensions are 3/4 in diameter by 1 in projection. This is a high quality rubber bumper at an affordable price from a trusted brand.
Catalog No. 50-276-5519