Learn More
Medchemexpress LLC HY-70063 10mg , Buparlisib CAS:944396-07-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY7006310MG
Encompass_Preferred
HY-70063 10mg Buparlisib CAS: 944396-07-0 Buparlisib (BKM120; NVP-BKM120) is a pan-class I PI3K inhibitor, with IC50s of 52, 166, 116 and 262 nM for p110α, p110β, p110δ and p110γ, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9277