Learn More
Cayman Chemical Buparvaquon 250mg
SDP
Supplier:  Cayman Chemical 21704250 MG
Encompass_Preferred
An inhibitor of cytochrome bc1 in the parasites T. annulata and T. parva (in vitro EC50 of 1.5 x 10-8 M and 6.1 x 10-10 M, respectively)
Catalog No. 50-298-6288