Learn More
Cayman Chemical Buphedron metabolItehydroc 5mg
SDP
Supplier:  Cayman Chemical 90014325 MG
Encompass_Preferred
A metabolite of buphedrone that features conversion of the β-keto group to β-hydroxy and is an enantiomeric mixture of the R,S and S,R orientations at carbons one and two, as in ephedrine
Catalog No. 50-289-4232