Learn More
Cayman Chemical BupIvacaIn 500mg
SDP
Supplier:  Cayman Chemical 16618500 MG
Encompass_Preferred
A sodium channel blocker and local anesthetic; inhibits sodium currents in rat dorsal horn neurons in a concentration-dependent manner; increases the firing threshold in rat sympathetic ganglia at 200 nM; blocks cardiac sodium channels in a use-dependent manner at 10 µM; inhibits respiration in cardiac cell mitochondria when palmitoyl-carnitine or acetyl-carnitine are used as substrates (IC50s = 0.78 and 0.37 mM, respectively); reduces thermal hyperplasia in a rat model of sciatic ligation injury when 0.6 ml of a 0.5% solution is administered into the perinerve space
Catalog No. 50-296-6324