Learn More
TARGETMOL CHEMICALS INC BUPIVACAINE 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T0030L50MG
Encompass_Preferred
Also available in 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Bupivacaine (AH-250) is a NMDA receptor inhibitor.Bupivacaine can block sodium L-calcium and potassium channels.Bupivacaine potently blocks SCN5A channels with the IC50 of 69.5 uM. Bupivacaine can be used for the research of chronic pain. purity: 100%
Catalog No. 50-258-1797