Learn More
Medchemexpress LLC Bupivacaine hydrochloride | 18010-40-7 | 99.85% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB0405A10MM/1ML
Encompass_Preferred
Bupivacaine hydrochloride is an NMDA receptor inhibitor. It blocks sodium, L-calcium, and potassium channels, and potently blocks SCN5A channels with an IC50 of 69.5 μM. This product is used for research related to chronic pain.
  • NMDA receptor inhibitor
  • Blocks sodium, L-calcium, and potassium channels
  • Potently blocks SCN5A channels with an IC50 of 69.5 μM
  • Used for chronic pain research
Catalog No. 50-004-08079