Learn More
Medchemexpress LLC Bupivacaine hydrochloride | 18010-40-7 | 99.99% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0405AR100MG
Encompass_Preferred
Bupivacaine hydrochloride is an analytical standard intended for research and analytical applications. It functions as an NMDA receptor inhibitor and can block sodium, L-calcium, and potassium channels, with potent blocking action on SCN5A channels. This compound can be used for research related to chronic pain.
  • Used as an analytical standard in qualitative, quantitative, and methodological research experiments.
  • Applications include HPLC, GC, and MS.
  • Inhibits NMDA receptors.
  • Blocks sodium, L-calcium, and potassium channels.
  • Potently blocks SCN5A channels.
  • Applicable for research into chronic pain.
  • Relevant for neuronal signaling, membrane transporter/ion channel research.
  • Used in reference standards for metabolism, drug testing, and human metabolites.
Catalog No. 50-004-05297