Learn More
American Radiolabeled Chemicals Inc BUPROPION 3H 50UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ART 175750UC
Encompass_Preferred
Bupropion-ring-3H 50uCi in ethanol, 1 mCi/ml, S.A. 10-20 Ci/mmol, M.W. 239.7 Shipped in Dry Ice, Exclusive
Catalog No. 50-901-104
This product is regulated by the federal government, and may not be ordered online.