Learn More
Apexbio Technology LLC Bupropion hydrochloride 31677-93-7 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B33265.1
Encompass_Preferred
Bupropion hydrochloride (CAS 31677-93-7) is a small-molecule aminoketone that acts as a dual inhibitor of norepinephrine and dopamine reuptake with IC values of 6 5 0 6 M and 3 4 0 4 M respectively It is widely utilized in research for its established roles in modulating monoaminergic neurotransmission associated with depression and smoking cessation In cellular studies bupropion hydrochloride has been shown to decrease cell viability in SH-SY5Y neuroblastoma cells selectively induce endoplasmic reticulum stress and activate JNK pathways leading to caspase 3-dependent apoptosis Additionally it has been reported to downregulate tyrosine hydroxylase expression impacting dopamine biosynthesis
Catalog No. 50-000-92435