Learn More
Medchemexpress LLC Bupropion morpholinol-d6 | 1216893-18-3 | 99.3% | 1 MG
SDP
Supplier:  Medchemexpress LLC HY12766S1MG
Encompass_Preferred
Bupropion morpholinol-d6 is the deuterated form of Bupropion morpholinol, a major metabolite of Bupropion. It inhibits Dopamine, Norepinephrine transporters and the α4β2 nicotinic receptor in vitro, contributing to antidepressant and smoking cessation activities.
  • Can be used as a tracer.
  • Suitable as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Inhibits dopamine and norepinephrine transporters.
  • Inhibits the α4β2 nicotinic receptor in vitro.
  • Contributes to antidepressant and smoking cessation activities.
  • For research use only.
Catalog No. 50-003-42186