Learn More
Med Vet International Miltex Friction Grip Diamond Bur, Flame Shaped, Extra Fine Grit, OgIVal, 1.0mm, German, Each, 860/010
Supplier:  Med Vet International MIL860/010
Encompass_Preferred
A safe end flame shaped diamond can be used for sub-gingival curettage and can remove sub-gingival calculus for initial root planning. Extra fine grit allows for more delicate procedures to be done. German manufactured steel provides the best in reliability and durability. 1.0mm.
Catalog No. 50-283-2035