Learn More
Wilmad Labglass BURET AUTO LAB GRADE 50ML
Supplier:  Wilmad Labglass LG4241T104
Encompass
Buret, Laboratory Grade, Automatic, with Straight PTFE Stopcock, Complete. Buret Capacity: 50mL; Subdivision: 0.1mL; Reservoir Capacity: 32 ounces; Stopcock Size: 2mm. Buret is filled by pressure supplied by the rubber bulb.
Catalog No. 50-804-295