Learn More
Enterprise Technology Solutions Business Source Masking Tape, 3" Core, 2"x60 Yds, 6 Rolls/PK, Natural Kraft
SDP
Supplier:  Enterprise Technology Solutions BSN16462PK
Encompass_Preferred
Utility-purpose, crepe paper-backed tape is designed for holding, bundling, sealing, noncritical masking and many other jobs where pressure-sensitive tape is needed. Design features a 3" core and approximate tensile strength of 8.5 kg. or 18.7 lb. per "
Catalog No. 50-008-32921