Learn More
Medchemexpress LLC Busulfan | 55-98-1 | 99.9% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB024510MM/1ML
Encompass_Preferred
Busulfan is a potent alkylating antineoplastic agent that causes DNA damage by cross-linking DNAs and DNA and proteins. It also inhibits thioredoxin reductase and induces apoptosis, acting as both an immunosuppressive and myeloablative chemotherapeutic agent.
  • Causes DNA damage by cross-linking DNAs and DNA and proteins
  • Inhibits thioredoxin reductase
  • Induces apoptosis
  • Functions as an immunosuppressive agent
  • Functions as a myeloablative chemotherapeutic agent
  • Can be used to induce aplastic anemia models
Catalog No. 50-004-06957