Learn More
Medchemexpress LLC Butanoic acid, 2,4-dihydroxy-3,3-dimethyl-, sodium salt (1:1), (2R)- | 60979-68-2 | MFCD29079001 | ≥95.0% | 170.14 g·mol⁻¹ | C6H11NaO4 | 25 MG
SDP
Supplier:  Medchemexpress LLC HYW57937725MG
Encompass_Preferred
Pantoic acid sodium is the sodium salt of pantoic acid used as a research reagent and an intermediate in the de novo biosynthesis of R-pantothenate (vitamin B5). It is supplied as a white to off-white solid with a molecular weight of 170.14 g·mol⁻¹ and a reported purity of ≥95.0%. Follow recommended storage: keep dry at room temperature; in solvent store at -80°C for long-term stability.
  • Intermediate in pantothenate biosynthesis
  • High purity ≥95.0%
  • White to off-white solid appearance
  • Supplied in small research quantities (25 mg)
  • Stable when stored under recommended conditions
Catalog No. 50-002-84236