Learn More
Medchemexpress LLC Butanoic acid, 2,4-dihydroxy-3,3-dimethyl-, sodium salt (2R)- | 60979-68-2 | MFCD29079001 | ≥95.0% | 170.14 | C6H11NaO4 | 5 MG

Supplier: Medchemexpress LLC HYW5793775MG
Pantoic acid sodium is the sodium salt of pantoic acid and an intermediate in the biosynthesis of R-pantothenate (vitamin B5). It is supplied as a white to off-white solid with established analytical characterization suitable for biochemical and biosynthetic research. The material is intended for research use only and is not for human or clinical applications.
- Intermediate in pantothenate (vitamin B5) biosynthesis studies.
- White to off-white solid, easy to handle.
- Purity ≥95.0% suitable for research applications.
- Molecular weight 170.14 and formula C6H11NaO4 for precise calculations.
- Store dry at room temperature; long-term stability in solvent at -80°C.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.