Learn More
Medchemexpress LLC Butanoic acid, 2-[[(9H-fluoren-9-ylmethoxy)carbonyl)amino]-, (2R)- | 170642-27-0 | MFCD00270336 | 325.36 | 50 G

Supplier: Medchemexpress LLC HYW00410150G
Fmoc-D-Abu-OH, also known as Fmoc-D-a-aminobutyric acid, is a high-purity, white to off-white solid. It serves as a biochemical assay reagent and is characterized by its specific molecular formula and weight.
- Purity of 98.1%
- Formula: C19H19NO4
- Molecular weight: 325.36
- Appearance: White to off-white solid
- Storage at room temperature for 3 years
- In solvent, store at -80°C for 2 years or -20°C for 1 year
- Used as a biochemical assay reagent
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.