Learn More
Medchemexpress LLC Butanoic acid, 4-[[(1,1-dimethylethoxy)carbonyl]amino]-2-[[(9h-fluoren-9-ylmethoxy)carbonyl]amino]-, (2r)- | 114360-56-4 | MFCD00798647 | 99.1% | 440.49 g/mol | C24H28N2O6 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0190325G
Encompass_Preferred
Fmoc-D-Dab(Boc)-OH is an Fmoc-protected D-2,4-diaminobutyric acid derivative used in Fmoc solid-phase peptide synthesis and in the preparation of modified peptides, including sequences investigated for antibacterial activity. The compound is supplied as a dry, high-purity material suitable for research-grade peptide assembly and analytical characterization.
  • High purity suitable for peptide synthesis and analysis.
  • Compatible with Fmoc solid-phase peptide synthesis workflows.
  • Protected side chain (Boc) for selective deprotection strategies.
  • Stable storage profile with multi-year shelf life at room temperature.
  • Available in multiple pack sizes to support varied research needs.
Catalog No. 50-003-01041