Learn More
Medchemexpress LLC Butanoic acid, 4-[4-fluoro-2-[1-[4-(1-methylethyl)phenyl]-5-phenyl-1H-pyrazol-3-yl]phenoxy]- | 2988135-14-2 | 99.9% | C28H27FN2O3 | 10MG
SDP
Supplier:  Medchemexpress LLC HY14599010MG
Encompass_Preferred
FABPs ligand 6 (MF6) is a research-grade small-molecule inhibitor of fatty acid-binding proteins FABP7 and FABP5. It is used in preclinical studies, including multiple sclerosis and mitochondrial function research, offering a well-characterized tool for biochemical and cellular assays.
  • High affinity for FABP7 (KD 20 nM) and FABP5 (KD 874 nM).
  • High purity (99.86%) suitable for biochemical assays.
  • Soluble in DMSO at high concentration (125 mg/mL) with ultrasonic assistance.
  • Solid off-white to light yellow appearance for easy handling.
  • Stable in solution when stored at -80°C for extended periods.
Catalog No. 50-002-59487