Learn More
Medchemexpress LLC Butanoic acid, 4-(acetylamino)- | 3025-96-5 | MFCD00020552 | 98.0% | 145.16 g/mol | C6H11NO3 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY101411500MG
Encompass_Preferred
4-Acetamidobutanoic acid (N-acetyl GABA) is a research-grade small molecule and a primary metabolite of gamma-aminobutyric acid (GABA). It has reported antioxidant and antibacterial activities and is intended for biochemical and pharmacological research, including metabolite profiling and receptor studies. The material is supplied as a high-purity solid suitable for analytical and preparative workflows.
  • High purity, 98.0%.
  • Molecular weight 145.16 g/mol; formula C6H11NO3.
  • Suitable for metabolite profiling and receptor pharmacology studies.
  • Available in solid quantities for analytical and preparative use.
  • COA and SDS available to support quality and safety documentation.
Catalog No. 50-002-47027