Learn More
Medchemexpress LLC Butanoic acid, 4-azido- | 54447-68-6 | MFCD11850695 | 99.5% | 129.12 g/mol | C4H7N3O2 | 5 G

Supplier: Medchemexpress LLC HYW5887045G
4-Azidobutyric acid is a small organic molecule bearing an azide functional group and a terminal carboxylic acid. It is used as a building block in chemical synthesis and bioconjugation, enabling click-chemistry reactions with terminal alkynes and coupling through the carboxyl group.
- Azide and carboxylic acid functionality for click chemistry and coupling
- High purity (99.48%) suitable for research applications
- Soluble in DMSO (≈200 mg/mL), facilitating solution preparation
- Available in multiple pack sizes, including 5 G for small-scale use
- Molecular weight 129.12 g/mol; molecular formula C4H7N3O2
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.