Learn More
Cayman Chemical ButenafIn13Cd3hydrochLR 10mg
SDP
Supplier:  Cayman Chemical 3401010 MG
Encompass_Preferred
An internal standard for the quantification of butenafine by GC- or LC-MS
Catalog No. 50-295-0182