Learn More
Cayman Chemical Butenoyl PAF 10mg
SDP
Supplier:  Cayman Chemical 6092910 MG
Encompass_Preferred
A product of the oxidative decomposition of 2-arachidonoyl phospholipids; 10-fold less potent than PAF as a PAF receptor agonist, but present in amounts 100-fold greater
Catalog No. 50-289-4631