Learn More
Selleck Chemical LLC Butoconazole nitrate S1833-50mg
SDP
Supplier:  Selleck Chemical LLC S183350MG
Encompass_Preferred
Butoconazole nitrate (RS-35887) is an anti-fungal agent that inhibits PHA-stimulated release of cytokines-- IL-2 TNF IFN and GM-CSF from human peripheral blood lymphocytes (IC50 of 7 2 g/mL 14 4 g/mL 7 36 g/mL and 7 6 g/mL respectively)
Catalog No. 50-310-9296