Learn More
Apexbio Technology LLC Butoconazole nitrate 64872-77-1 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B19025.1
Encompass_Preferred
Butoconazole nitrate (CAS 64872-77-1) is a small-molecule inhibitor designed to disrupt fungal sterol biosynthesis thereby impacting fungal cell membrane integrity Butoconazole nitrate exerts its biological activity primarily through inhibition of fungal sterol (ergosterol) production In laboratory studies butoconazole nitrate demonstrates inhibitory effects against Candida species especially Candida albicans with an IC50 in the low micromolar range Based on these pharmacological properties butoconazole nitrate holds research potential in studies on fungal pathogenesis antifungal resistance mechanisms and in vitro fungal infection models
Catalog No. 50-000-89859