Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YMC America, Inc BUTYL 5UM 300A 250X20MM
SDP

Supplier:  YMC America, Inc BU30S052520WT

Encompass_Preferred

Butyl 5Um; 300A 250 X 20mm

Catalog No. 50-900-05130


May include imposed supplier surcharges.
Only null left
Add to Cart
Request a Quote

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.