Learn More
Medchemexpress LLC Butyl isobutyl phthalate | 17851-53-5 | 1 ML

Supplier: Medchemexpress LLC HYN737710MM/1ML
Butyl isobutyl phthalate is a chemical compound isolated from the rhizoid of Laminaria japonica. It acts as a non-competitive α-glucosidase inhibitor with an IC₅₀ value of 38 μM. This compound has demonstrated a hypoglycemic effect, indicating its potential for use in diabetes treatment research.
- Isolated from the rhizoid of Laminaria japonica.
- Functions as a non-competitive α-glucosidase inhibitor.
- Exhibits a hypoglycemic effect.
- Shows potential for diabetes treatment.
- Has an IC₅₀ value of 38 μM against α-glucosidase.
- Molecular formula is C₁₆H₂₂O₄.
- Molecular weight is 278.34.
- Targets glucosidase.
- Purity of 99.31%.
- Appearance is a colorless to light yellow liquid.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.