Learn More
Sigma Aldrich Fine Chemicals Biosciences Butyl SepharoseR 4 Fast Fl
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GE17098001
Encompass
Butyl Sepharose(R) 4 Fast FLow is a well established standard aliphatic hydrophobic interaction chromatography (HIC) medium for capture and intermediate purification of larger proteins.
Catalog No. 50-290-8395