Learn More
Medchemexpress LLC Butylated hydroxytoluene-d21 | 64502-99-4 | 98.7% | 241.48 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYY0172S
Encompass_Preferred
Butylated hydroxytoluene-d21 is a deuterium-labeled version of Butylated hydroxytoluene, functioning as an antioxidant commonly found in food and food-related products. It also acts as a Ferroptosis inhibitor. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules as tracers for quantitative analysis during drug development, potentially influencing pharmacokinetic and metabolic profiles.
  • Can be utilized as a tracer
  • Suitable for use as an internal standard in quantitative analysis methods such as NMR, GC-MS, or LC-MS
Catalog No. 50-003-65896