Learn More
Medchemexpress LLC Butylated hydroxytoluene-d3 | 86819-59-2 | 99.2% | 223.37 | 5 MG
SDP
Supplier:  Medchemexpress LLC 5MGHYY0172S2
Encompass_Preferred
Butylated hydroxytoluene-d3 is the deuterium-labeled Butylated hydroxytoluene. It is an antioxidant widely used in foods and food-related products, and also functions as a Ferroptosis inhibitor. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration, involving stable heavy isotopes, is used in drug development to quantitate and potentially affect pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-93373