Learn More
Medchemexpress LLC Butylated hydroxytoluene-d3 | 86819-59-2 | MFCD14706649 | 99.2% | 223.37 | C15H21D3O | 100mg
SDP
Supplier:  Medchemexpress LLC HYY0172S2100MG
Encompass_Preferred
Butylated hydroxytoluene-d3 is the deuterium labeled Butylated hydroxytoluene[1] Butylated hydroxytoluene is an antioxidant widely used in foods and in food-related products[2] Butylated hydroxytoluene is a Ferroptosis inhibitor[3]
Catalog No. 50-002-69007