Learn More
Strem, An Ascensus Company CAS# 321921-71-5. 1g. Butyldi-1-adamantylphosphine, min. 95% [cataCXium A]. MFCD05861606
SDP
Supplier:  Strem, An Ascensus Company 1504831G
Encompass
CAS# 321921-71-5. 1g. Butyldi-1-adamantylphosphine, min. 95% [cataCXium A]. MFCD05861606. Molecular Weight: 358.54. Molecular Formula: C4H9(C10H15)2P. Color/form: white to yellow pwdr. Strem# 15-0483. http://www.strem.com/catalog/v/15-0483/
Catalog No. 50-901-09921