Learn More
Selleck Chemical LLC Butylparaben S4584-100mg
SDP
Supplier:  Selleck Chemical LLC S4584100MG
Encompass_Preferred
Butylparaben (Butyl parahydroxybenzoate Butyl 4-hydroxybenzoate) is a chemical compound commonly used as an antifungal preservative in cosmetic products
Catalog No. 50-311-0314