Learn More
Medchemexpress LLC Butylparaben-13C6 | 1416711-53-9 | 99.75% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB1431S11MG
Encompass_Preferred
Butylparaben-13C6 is the 13C labeled version of Butylparaben, an organic compound known for its effectiveness as an antimicrobial preservative in cosmetics and its use in medication suspensions and as a flavoring additive in food. This compound is used as a tracer and an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes of hydrogen, carbon, and other elements
  • Potential to affect the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-13693