Learn More
Medchemexpress LLC Butylparaben | 94-26-8 | 98.9% | 10 G
SDP
Supplier:  Medchemexpress LLC HYB143110G
Encompass_Preferred
Butylparaben is an organic compound that serves as an effective antimicrobial preservative. It is utilized in various applications, including cosmetics, active molecule suspensions, and as a food flavoring additive.
  • Successful antimicrobial preservative
  • Can be used in cosmetics
  • Suitable for active molecule suspensions
  • Functions as a food flavoring additive
Catalog No. 50-004-12911