Learn More
Medchemexpress LLC Butylparaben | 94-26-8 | 98.9% | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYB1431
Encompass_Preferred
Butylparaben is an organic compound that serves as an antimicrobial preservative in cosmetics, active molecule suspensions, and as a food flavoring additive. It can also be used in vitro to inhibit porcine oocyte fertilization, cleavage, and blastocyst formation, and in vivo studies have shown no systemic toxicity at certain doses.
  • Serves as an antimicrobial preservative
  • Can be used in cosmetics and active molecule suspensions
  • Functions as a food flavoring additive
  • Inhibits porcine oocyte fertilization and development in vitro
  • Induces DNA damage, apoptosis, and autophagy in porcine oocytes in vitro
  • Shows no systemic toxicity in rats at specific doses in vivo
Catalog No. 50-004-14221