Learn More
Medchemexpress LLC Butylparaben-d4 | 1219798-67-0 | 98.6% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB1431S1MG
Encompass_Preferred
Butylparaben-d4 is the deuterium labeled Butylparaben, an organic compound that has been successfully used as an antimicrobial preservative in cosmetics, medication suspensions, and as a flavoring additive in food.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes into drug molecules for quantitation during drug development.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Classified as an isotope-labeled compound and an internal standard for food.
Catalog No. 50-004-13505