Learn More
Medchemexpress LLC Butylparaben-d9 | 1216904-65-2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB1431S25MG
Encompass_Preferred
Butylparaben-d9 (Butyl parahydroxybenzoate-d9; Butyl paraben-d9) is a deuterium-labeled form of Butylparaben (HY-B1431). Deuterium labeling involves incorporating stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules, primarily for use as tracers in quantitative analysis during drug development. This process has garnered attention for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer in drug development processes.
  • Can be utilized as an internal standard for quantitative analysis.
  • Compatible with analytical techniques such as NMR, GC-MS, and LC-MS.
  • Deuterium labeling can impact pharmacokinetic profiles of drugs.
  • Deuterium labeling can affect metabolic profiles of drugs.
  • Stable heavy isotopes for reliable research applications.
  • For research use only.
Catalog No. 50-004-13706