Learn More
Medchemexpress LLC Butylparaben (Standard) | 94-26-8 | 98.9% | 250 MG

Supplier: Medchemexpress LLC 250MGHYB1431R
Butylparaben (Standard) is the analytical standard of Butylparaben. It is an organic compound used as an antimicrobial preservative in cosmetics, in active molecule suspensions, and as a food flavoring additive. It is intended for research and analytical applications.
- Analytical standard for Butylparaben.
- Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
- Appearance: Solid, white to off-white color.
- Molecular weight: 194.23.
- Formula: C₁₁H₁₄O₃.
- Structure classification: Phenols, Monophenols.
- Initial source: Endogenous metabolite.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.