Learn More
Medchemexpress LLC Butylparaben (Standard) | 94-26-8 | 98.9% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB1431R50MG
Encompass_Preferred
Butylparaben (Standard) is an analytical standard intended for research and analytical applications. This organic compound acts as a successful antimicrobial preservative in cosmetics and can also be used in active molecule suspensions and as a food flavoring additive.
  • Used as an analytical standard and reference standard assay
  • Employed in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
  • Functions as an antimicrobial preservative in cosmetics
  • Can be used in active molecule suspensions
  • Serves as a food flavoring additive
Catalog No. 50-004-12605