Learn More
Medchemexpress LLC Butyramide | 541-35-5 | MFCD00041894 | 98.1% | 87.12 g·mol⁻¹ | C4H9NO | 25 G

Supplier: Medchemexpress LLC 25GHYW088319
Butyramide is a small-molecule amide used as a biochemical reagent in life-science research. It is supplied as a powder for laboratory and analytical applications.
- Intended for research use only.
- CAS number 541-35-5.
- Molecular formula C4H9NO.
- Molecular weight 87.12 g·mol⁻¹.
- Typical purity about 98.05% as supplied.
- Available in a 25 G pack size.
- Powder storage: -20 °C for long-term, 4 °C for shorter-term storage.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.