Learn More
Medchemexpress LLC Butyrophenone | 495-40-9 | 99.0% | 148.20 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01026510G
Encompass_Preferred
Butyrophenone, also known as 1-Phenylbutan-1-one, is a dopamine receptor (DA Receptor) antagonist primarily used in research contexts, particularly for studies on mental disorders. This chemical compound is available in a liquid form, presenting as colorless to light yellow. It is highly soluble in DMSO, making it suitable for various in vitro applications.
  • Suitable for research on mental disorders
  • Functions as a dopamine receptor antagonist
  • Available in a liquid, colorless to light yellow form
  • Soluble in DMSO for in vitro experiments
  • Can be stored at room temperature for extended periods
Catalog No. 50-004-53986